Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anza™ Alkaline Phosphatase

Catalog No. p-6543163
Encompass_Preferred
Change view
Click to view available options
Quantity:
2000 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN2208 2000 Reactions
IVGN2204 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. IVGN2208 Supplier Invitrogen™ Supplier No. IVGN2208
Only null left
Add to Cart
Add to Cart

Includes

Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• Utilizes the Anza 10X Buffer
• One-step digestion and dephosphorylation protocol
• Two-step dephosphorylated protocols available
• Heat inactivated in 5 minutes

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 mL Anza Alkaline Phosphatase
1 mL Anza Buffer (10X)
1 mL Anza Red Buffer (10X)

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme Alkaline Phosphatase
Compatible Buffer 10X Buffer
Quantity 2000 Reactions
Product Type Alkaline Phosphatase
Product Line Anza

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.