Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP-2 gamma Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB318396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318396 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318396 Supplier Novus Biologicals Supplier No. NBP298897JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP-2 gamma Polyclonal antibody specifically detects AP-2 gamma in Human samples. It is validated for Western Blot

Specifications

Antigen AP-2 gamma
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Activating enhancer-binding protein 2 gamma, AP2-GAMMA, ERF1, estrogen receptor factor 1, hAP-2g, TFAP2G, transcription factor AP-2 gamma, transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma), transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma), Transcription factor ERF-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AP-2 gamma.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7022
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.