Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP-2 gamma Antibody, Novus Biologicals™
SDP

Catalog No. NB1001516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001516 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001516 Supplier Novus Biologicals Supplier No. NB1001516
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AP-2 gamma Polyclonal specifically detects AP-2 gamma in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.

Specifications

Antigen AP-2 gamma
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2 - 3 ug/ml, Immunohistochemistry-Paraffin 2 - 3 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q92754
Gene Alias Activating enhancer-binding protein 2 gamma, AP2-GAMMA, ERF1, estrogen receptor factor 1, hAP-2g, TFAP2G, transcription factor AP-2 gamma, transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma), transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma), Transcription factor ERF-1
Gene Symbols TFAP2C
Host Species Goat
Immunogen Peptide with sequence C-NKTLEKMEKHRK corresponding to C-Terminus according to NP_003213.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7022
Test Specificity Cross reacts with Human. Expected from sequence similarity: Rat, Mouse, Dog, Pig and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.