Learn More
AP1S1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | AP1S1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0319
![]() |
Bethyl Laboratories, Inc
A305396AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
AP1S1 is part of the clathrin coat assembly complex which links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. This protein, as well as beta-prime-adaptin, gamma-adaptin, and the medium (mu) chain AP47, form the AP-1 assembly protein complex located at the Golgi vesicle.Specifications
AP1S1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
AP19, CLAPS1, SIGMA1A, WUGSC:H_DJ0747G18.2, AP-1 complex subunit sigma-1A, HA1 19 kDa subunit, adapter-related protein complex 1 sigma-1A subunit, adaptor protein complex AP-1 sigma-1A subunit, clathrin assembly protein complex 1 sigma-1A small chain, clathrin coat assembly protein AP19, clathrin-associa | |
Between 108 and 158 | |
Primary | |
4° C | |
AP1S1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P61966, P61967 | |
1174, 11769 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.