Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Antibody (3B5), Novus Biologicals™
SDP

Catalog No. NB10074359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074359 Supplier Novus Biologicals Supplier No. NB10074359
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

AP2 alpha Monoclonal specifically detects AP2 alpha in Chicken, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen AP2 alpha
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3B5
Conjugate Unconjugated
Dilution Western Blot 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:100 - 1:250, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50
Gene Accession No. P05549
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Gene Symbols TFAP2A
Host Species Mouse
Immunogen N-terminus of human AP2a.
Molecular Weight of Antigen 48 kDa
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7020
Test Specificity AP-2 (3B5)
Target Species Chicken, Human, Chicken, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.