Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Antibody (A6/2/2), PE, Novus Biologicals™
SDP

Catalog No. NB600202PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600202PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600202PE Supplier Novus Biologicals Supplier No. NB600202PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen AP2 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone A6/2/2
Conjugate PE
Dilution Flow Cytometry
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Gene Symbols TFAP2A
Host Species Mouse
Immunogen Peptide to C-Terminal of AP2 alpha coupled to KLH. PNSHTDNNAKSSDKEEKHRK
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7020
Test Specificity This antibody detects both the alpha and beta forms of AP2.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.