Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Antibody (AP2a 8G8/5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N102639A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102639A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102639A532 Supplier Novus Biologicals Supplier No. NB1002639AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for ChIP Assay, Immunohistochemistry, Western Blot.

Specifications

Antigen AP2 alpha
Applications ChIP Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AP2a 8G8/5
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Gene Symbols TFAP2A
Host Species Mouse
Immunogen Bacterialy expressed AP-2a/AP-2g fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7020.0
Test Specificity AP2a 8G8/5 is specific for the alpha form of AP-2.Antigen recovery using microwave or pressure cooking required for paraffin embedded tissue.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.