Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Antibody, Novus Biologicals™
SDP

Catalog No. NB10056332T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056332T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056332T Supplier Novus Biologicals Supplier No. NB10056332SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP2 alpha Polyclonal specifically detects AP2 alpha in Bovine, Canine, Human, Mouse, Primate, Rat, Sheep, Xenopus species samples. It is validated for Dot Blot, Western Blot.

Specifications

Antigen AP2 alpha
Applications Dot Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Dot Blot
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_001035890
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Gene Symbols TFAP2A
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human AP-2 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7020
Test Specificity The amino acid sequence used as immunogen has 100% identity in human, mouse, rat, bovine, dog, monkey, sheep and Xenopus, and 93% in chicken.
Target Species Bovine, Canine, Rat, Human, Mouse, Primate, Rat, Sheep, Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.