Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Rabbit anti-Human, Mouse, Clone: 8I8E4, Novus Biologicals™
SDP

Catalog No. NB167308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167308 20 μg
NB167307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167308 Supplier Novus Biologicals Supplier No. NBP31610820UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen AP2 alpha
Applications Western Blot
Classification Monoclonal
Clone 8I8E4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human AP2 alpha (P05549).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7020
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.