Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP2 alpha Rabbit anti-Human, Mouse, Clone: 8I8E4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | AP2 alpha |
---|---|
Clone | 8I8E4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human, Mouse samples. It is validated for Western BlotSpecifications
AP2 alpha | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7020 | |
IgG | |
Affinity purified |
8I8E4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human AP2 alpha (P05549). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title