Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2M1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132272 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132272 Supplier Novus Biologicals Supplier No. NBP132272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP2M1 Polyclonal specifically detects AP2M1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AP2M1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.57mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
Gene Alias Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein
Gene Symbols AP2M1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AP2M1. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1173
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.