Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2M1 Antibody (OTI1E9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270417R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270417R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270417R Supplier Novus Biologicals Supplier No. NBP270417R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AP2M1 Monoclonal specifically detects AP2M1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AP2M1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1E9
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein
Gene Symbols AP2M1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 97-383 of human AP2M1 (NP_001020376) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1173.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.