Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2S1 Rabbit anti-Mouse, Clone: 4G1G1, Novus Biologicals™
SDP

Catalog No. NB168683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168683 100 μg
NB168684 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168683 Supplier Novus Biologicals Supplier No. NBP316796100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AP2S1 Monoclonal antibody specifically detects AP2S1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen AP2S1
Applications Western Blot
Classification Monoclonal
Clone 4G1G1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Adapter-related protein complex 2 sigma subunit, Adaptor protein complex AP-2 subunit sigma, adaptor-related protein complex 2, sigma 1 subunit, AP17HA2 17 kDa subunit, AP-2 complex subunit sigma, CLAPS2AP17-DELTA, Clathrin assembly protein 2 small chain, Clathrin coat assembly protein AP17, Clathrin coat-associated protein AP17, clathrin-associated/assembly/adaptor protein, small 2 (17kD), Plasma membrane adaptor AP-2 17 kDa protein, Sigma2-adaptin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human AP2S1 (P53680).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1175
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.