Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP3B1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16892720UL
Description
AP3B1 Polyclonal specifically detects AP3B1 in Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
AP3B1 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Adapter-related protein complex 3 subunit beta-1, Adaptor protein complex AP-3 subunit beta-1, adaptor-related protein complex 3, beta 1 subunit, ADTB3, ADTB3APE, AP-3 complex subunit beta-1, beta-3A-adaptin, beta3A-adaptin, Clathrin assembly protein complex 3 beta-1 large chain, HPS, HPS2AP-3 complex beta-3A subunit | |
Rabbit | |
121 kDa | |
20 μL | |
Primary | |
This product is specific to AP3B1. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
AP3B1 | |
Synthetic peptides corresponding to Ap3b1 (adaptor-related protein complex 3, beta 1 subunit) The peptide sequence was selected from the N terminal of Ap3b1. Peptide sequence MSSNSFAYNEQSGGGEAAELGQEATSTISSSGAFGLFSSDWKKNEDLKQM. | |
Affinity Purified | |
RUO | |
8546 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction