Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3B1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen AP3B1
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168927
SDP
View Documents
Novus Biologicals
NBP168927
100 μL
Each for $487.50
Only null left
Add to Cart
 
NBP16892720
SDP
View Documents
Novus Biologicals
NBP16892720UL
20 μL N/A N/A N/A
Description

Description

AP3B1 Polyclonal specifically detects AP3B1 in Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

AP3B1
Unconjugated
RUO
8546
Synthetic peptides corresponding to Ap3b1 (adaptor-related protein complex 3, beta 1 subunit) The peptide sequence was selected from the N terminal of Ap3b1. Peptide sequence MSSNSFAYNEQSGGGEAAELGQEATSTISSSGAFGLFSSDWKKNEDLKQM.
Primary
121 kDa
Polyclonal
Rabbit
Adapter-related protein complex 3 subunit beta-1, Adaptor protein complex AP-3 subunit beta-1, adaptor-related protein complex 3, beta 1 subunit, ADTB3, ADTB3APE, AP-3 complex subunit beta-1, beta-3A-adaptin, beta3A-adaptin, Clathrin assembly protein complex 3 beta-1 large chain, HPS, HPS2AP-3 complex beta-3A subunit
AP3B1
IgG
This product is specific to AP3B1.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.