Learn More
Invitrogen™ AP3B2 Polyclonal Antibody

Description
Immunogen sequence: APVFMSENEF KKEQGKLMGM NEITEKLMLP DTCRSDHIVV QKVTATANLG RVPCGTSDEY RFAGRTLTGG SLVLLTLDAR PAGAAQLTVN SEKMVIGTML VKDVIQ Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 93%.
Specifications
Specifications
| Antigen | AP3B2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AP3B2 |
| Gene Accession No. | Q13367 |
| Gene Alias | [b]-NAP; adapter-related protein complex 3 subunit beta-2; adaptor protein complex AP-3 beta-2 subunit; adaptor protein complex AP-3 subunit beta-2; adaptor related protein complex 3 beta 2 subunit; adaptor related protein complex 3, beta 2 subunit; adaptor-related protein complex 3 subunit beta-2; adaptor-related protein complex 3, beta 2 subunit; adaptor-related protein complex AP-3, beta 2 subunit; AI549966; ap3; AP-3 complex beta-2 subunit; AP-3 complex subunit beta-2; Ap3b2; AU042881; beta3B; beta-3B-adaptin; beta3B-adaptin; beta-NAP; Clathrin a; clathrin assembly protein complex 3 beta-2 large chain; NAPTB; Neuronal adaptin-like protein, beta-subunit; neuron-specific vesicle coat protein beta-NAP |
| Gene Symbols | AP3B2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.