Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3M1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227068 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227068 Supplier Novus Biologicals Supplier No. NBP227068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP3M1 Polyclonal specifically detects AP3M1 in Human, Bovine, Canine, Chicken, Chinese Hamster, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AP3M1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y2T2
Gene Alias adaptor-related protein complex 3, mu 1 subunit, AP-3 adapter complex mu3A subunit, MGC22164, mu-3A subunit, mu3A-adaptin
Gene Symbols AP3M1
Host Species Rabbit
Immunogen The immunogen for this antibody was a portion of amino acid 140-190 of human AP3M1.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26985
Test Specificity The amino acid sequence used as immunogen for the AP3 antibody is 100% homologous in human, rhesus monkey, wild pig, dog, cow, horse, opossum, orangutan, mouse, rat, wild turkey, zebrafinch, chicken, new world monkey, Chinese hamster, macaque, rabbit, Carolina anole and platypus.
Target Species Human, Bovine, Canine, Chicken, Chinese Hamster, Equine, Other
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.