Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3S1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15544920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15544920 20 μL
NBP155449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15544920 Supplier Novus Biologicals Supplier No. NBP15544920UL

Rabbit Polyclonal Antibody

AP3S1 Polyclonal specifically detects AP3S1 in Human samples. It is validated for Western Blot.

Specifications

Antigen AP3S1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q92572
Gene Alias Adapter-related protein complex 3 sigma-1 subunit, adaptor-related protein complex 3, sigma 1 subunit, AP-3 complex sigma-3A subunit, AP-3 complex subunit sigma-3A, CLAPS3AP-3 complex subunit sigma-1, Clathrin-associated/assembly/adapter protein, small 3, clathrin-associated/assembly/adaptor protein, small 3 (22kD), Sigma3A, sigma-3A-adaptin, Sigma3A-adaptin, Sigma-adaptin 3a
Gene Symbols AP3S1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to AP3S1(adaptor-related protein complex 3, sigma 1 subunit) The peptide sequence was selected from the N terminal of AP3S1 (NP_001275). Peptide sequence IKAILIFNNHGKPRLSKFYQPYSEDTQQQIIRETFHLVSKRDENVCNFLE.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1176
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.