Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APAAP Complex Antibody (WR31), Novus Biologicals™
SDP

Catalog No. NB10062212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062212 0.5 mL
NB017444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062212 Supplier Novus Biologicals Supplier No. NB10062212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APAAP Complex Monoclonal specifically detects APAAP Complex in Non-species specific samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen APAAP Complex
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone WR31
Conjugate Unconjugated
Dilution Immunohistochemistry 1:30-1:50, Immunohistochemistry-Paraffin 1:30-1:50, Immunohistochemistry-Frozen 1:30-1:50
Host Species Mouse
Immunogen Bovine alkaline phosphatase
Purification Method Protein A or G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Embryonic Stem Cell Markers
Primary or Secondary Primary
Test Specificity APAAP complexes are designed for use as detection reagents in immunohistochemistry procedures. They are used in combination with a suitable bridging antibody and appropriate visualization system.
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.