Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APAF-1 Antibody (18H2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB173316 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173316 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173316 Supplier Novus Biologicals Supplier No. NBP280094JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

APAF-1 Monoclonal antibody specifically detects APAF-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen APAF-1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 18H2
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413
Host Species Rat
Immunogen Recombinant mouse APAF-1 (aa 1-97) containing the N-terminal CARD domain.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 317
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.