Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBA1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB344201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344201 Supplier Novus Biologicals Supplier No. NBP241085MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APBA1 Polyclonal antibody specifically detects APBA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen APBA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Adapter protein X11alpha, amyloid beta (A4) precursor protein-binding, family A, member 1, amyloid beta A4 precursor protein-binding family A member 1, D9S411Eadaptor protein X11alpha, LIN10, Mint-1, MINT1amyloid beta (A4) precursor protein-binding, family A, member 1 (X11), Neuronal Munc18-1-interacting protein 1, Neuron-specific X11 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, UROP11, X11A, X11X11ALPHA
Host Species Rabbit
Immunogen Antibody was raised against a 12 amino acid synthetic peptide near the center of the mouse UROP11. The immunogen is located within the first 50 amino acids of UROP11. Amino Acid Squence: FHLPRYRNKAKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Vision
Primary or Secondary Primary
Gene ID (Entrez) 320
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.