Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | APBA1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
APBA1 Polyclonal specifically detects APBA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
APBA1 | |
Polyclonal | |
Rabbit | |
Membrane Trafficking and Chaperones, Vision | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
320 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ALPNHLHFHSLEHEEAMNAAYSGYVYTHRLFHRGEDEPYSEPYADYGGLQEHVYEEIGDAPELDARDGLRLYEQERDE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
Adapter protein X11alpha, amyloid beta (A4) precursor protein-binding, family A, member 1, amyloid beta A4 precursor protein-binding family A member 1, D9S411Eadaptor protein X11alpha, LIN10, Mint-1, MINT1amyloid beta (A4) precursor protein-binding, family A, member 1 (X11), Neuronal Munc18-1-interacting protein 1, Neuron-specific X11 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, UROP11, X11A, X11X11ALPHA | |
APBA1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title