Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBA2 Antibody (1C12A4) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP261881

 View more versions of this product

Catalog No. NBP2-61881


Only null left
Add to Cart

Description

Description

APBA2 Monoclonal specifically detects APBA2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

APBA2
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200
APBA2
Purified recombinant fragment of human APBA2 (AA: 15-158) expressed in E. Coli.
Protein G purified
Primary
Human
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1C12A4
Unconjugated
PBS with 0.05% Sodium Azide
Mouse
63.4 kDa
0.1 mL
321
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.