Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA2 Antibody (1C12A4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | APBA2 |
---|---|
Clone | 1C12A4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61881
![]() |
Novus Biologicals
NBP261881 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
APBA2 Monoclonal specifically detects APBA2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
APBA2 | |
1 mg/mL | |
Monoclonal | |
Purified | |
321 | |
Purified recombinant fragment of human APBA2 (AA: 15-158) expressed in E. Coli. | |
Primary | |
63.4 kDa |
1C12A4 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Unconjugated | |
Mouse | |
APBA2 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title