Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA2 Antibody (7G5G11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261880
Description
APBA2 Monoclonal specifically detects APBA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
APBA2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
APBA2 | |
Purified recombinant fragment of human APBA2 (AA: 15-158) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
7G5G11 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
Mouse | |
122.4 kDa | |
0.1 mL | |
321 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction