Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA2 Antibody (7G5G11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | APBA2 |
---|---|
Clone | 7G5G11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61880
![]() |
Novus Biologicals
NBP261880 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
APBA2 Monoclonal specifically detects APBA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
APBA2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
APBA2 | |
IgG1 | |
Protein G purified |
7G5G11 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
Monoclonal | |
Purified | |
321 | |
Purified recombinant fragment of human APBA2 (AA: 15-158) expressed in E. Coli. | |
Primary | |
122.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title