Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBA3/Mint-3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB327304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327304 Supplier Novus Biologicals Supplier No. NBP306040APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APBA3/Mint-3 Polyclonal antibody specifically detects APBA3/Mint-3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen APBA3/Mint-3
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Adapter protein X11gamma, amyloid beta (A4) precursor protein-binding, family A, member 3, amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2), MGC:15815, mint3, Mint-3, Neuronal Munc18-1-interacting protein 3, Neuron-specific X11L2 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, X11L2amyloid beta A4 precursor protein-binding family A member 3, X11-like 2 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human APBA3/Mint-3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Vision
Primary or Secondary Primary
Gene ID (Entrez) 9546
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.