Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Antibody (5F2), Novus Biologicals™
SDP

Catalog No. NB297967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297967 100 μL
NB297966 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297967 Supplier Novus Biologicals Supplier No. NBP326505100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

APC Monoclonal antibody specifically detects APC in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen APC
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5F2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC
Host Species Rabbit
Immunogen A synthesized peptide derived from Human APC [UniProt P25054]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 324
Target Species Human, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.