Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Antibody (ALi 12-28), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB383605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383605 Supplier Novus Biologicals Supplier No. NBP308922C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APC Monoclonal antibody specifically detects APC in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen APC
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone ALi 12-28
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC
Host Species Mouse
Immunogen Recombinant fragment of human APC protein (around aa 1-433) (Uniprot: P25054)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 324
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.