Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC-Cy7 Mouse Anti-Human CD29 (Integrin β1), BD Pharmingen™

Catalog No. BDB569865 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569865 100 Tests
BDB569941 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569865 Supplier BD Biosciences Supplier No. 569865
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD29 (Integrin β1) Monoclonal antibody specifically detects CD29 (Integrin β1) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29 (Integrin β1)
Applications Flow Cytometry
Classification Monoclonal
Clone TS2/16
Conjugate APC-Cyanine7
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias FNRB; GPIIA; ITGB1; MDF2; MSK12; VLA-BETA; VLAB; glycoprotein IIa
Host Species Mouse
Immunogen Human CTL Cell Line
Quantity 100 Tests
Regulatory Status RUO
Gene ID (Entrez) 3688
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.