Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC-PCI Complex Antibody (M36), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP241380A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP241380A94 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP241380A94 Supplier Novus Biologicals Supplier No. NBP241380AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APC-PCI Complex Monoclonal specifically detects APC-PCI Complex in Human samples. It is validated for ELISA.

Specifications

Antigen APC-PCI Complex
Applications ELISA
Classification Monoclonal
Clone M36
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%).
Purification Method Protein A or G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Antibody is specific for APC:PCI complex and cleaved PCI but has little reactivity towards native PCI. The dissociation constants (as determined by standard surface plasmon resonance technique) are: Kd= 4x10e-10 M for PCI in complex with APC and 2x10e-10 M for cleaved PCI. The value of Kd for native PCI is too low to measure, i.e. > 10e-5 M.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.