Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC-PCI Complex Antibody (M36), PerCP, Novus Biologicals™
SDP

Catalog No. NB343284 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343284 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343284 Supplier Novus Biologicals Supplier No. NBP241380PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APC-PCI Complex Monoclonal antibody specifically detects APC-PCI Complex in Human samples. It is validated for ELISA

Specifications

Antigen APC-PCI Complex
Applications ELISA
Classification Monoclonal
Clone M36
Conjugate PerCP
Dilution ELISA
Formulation PBS
Host Species Mouse
Immunogen Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.