Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc1 Antibody (7G9C7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61887 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61887 Supplier Novus Biologicals Supplier No. NBP261887
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apc1 Monoclonal specifically detects Apc1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Apc1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 7G9C7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR
Gene Symbols ANAPC1
Host Species Mouse
Immunogen Purified recombinant fragment of human ANAPC1 (AA: 12-155) expressed in E. Coli.
Molecular Weight of Antigen 216 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64682
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.