Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10086985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10086985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10086985 Supplier Novus Biologicals Supplier No. NB10086985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Apc1 Polyclonal specifically detects Apc1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Apc1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9H1A4
Gene Alias anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR
Gene Symbols ANAPC1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human anaphase promoting complex subunit 1 using the numbering given in entry NP_073153.1 (GeneID 64682).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64682
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.