Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc1, p Ser377 Antibody, Novus Biologicals™
SDP

Catalog No. NB600895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600895 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600895 Supplier Novus Biologicals Supplier No. NB600895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apc1 Polyclonal specifically detects Apc1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Apc1 (p Ser377)
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 5.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9H1A4
Gene Alias anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR
Gene Symbols ANAPC1
Host Species Rabbit
Immunogen This affinity purified Apc1 [p Ser377] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region surrounding pS377 of Human Apc1 protein. (Uniprot: Q9H1A4)
Molecular Weight of Antigen 216.5 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 64682
Test Specificity This product is an affinity purified antibody produced by immunoaffinity chromatography using phospho peptide coupled to agarose beads followed by solid phase adsorption(s) against non-phospho peptide and non-specific peptide to remove any unwanted reactivities. This antibody is specific for phosphorylated human Apc1 protein at the pS377 residue. A BLAST analysis was used to suggest reactivity with this protein from human, dog, rat, and bovine based on 100% homology for the immunogen sequence. Cross reactivity with Apc1 protein from mouse is expected, as this sequence show 90% homologous between human and mouse. Cross reactivity with Apc1 homologues from other sources has not been determined. Minimal reactivity is expected with the non-phosphorylated form of the protein.
Target Species Human, Rat, Bovine, Canine
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.