Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc10 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356422 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356422 Supplier Novus Biologicals Supplier No. NBP177157MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apc10 Polyclonal antibody specifically detects Apc10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Apc10
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias anaphase promoting complex subunit 10, anaphase-promoting complex subunit 10, APC10DOC1, Cyclosome subunit 10, DKFZP564L0562
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human APC10. The immunogen is located within amino acids 60 - 110 of APC10. Amino Acid Squence: IYADYKSDESYTPSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10393
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.