Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158197
Description
Apc10 Polyclonal specifically detects Apc10 in Human samples. It is validated for Western Blot.Specifications
Apc10 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
anaphase promoting complex subunit 10, anaphase-promoting complex subunit 10, APC10DOC1, Cyclosome subunit 10, DKFZP564L0562 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
This product is specific to Subunit or Isoform: 10. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9UM13 | |
ANAPC10 | |
Synthetic peptides corresponding to ANAPC10(anaphase promoting complex subunit 10) The peptide sequence was selected from the N terminal of ANAPC10. Peptide sequence MTTPNKTPPGADPKQLERTGTVREIGSQAVWSLSSCKPGFGVDQLRDDNL. | |
100 μL | |
Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
10393 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction