Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Apc10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Apc10 Polyclonal specifically detects Apc10 in Human samples. It is validated for Western Blot.Specifications
Apc10 | |
Polyclonal | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
anaphase promoting complex subunit 10, anaphase-promoting complex subunit 10, APC10DOC1, Cyclosome subunit 10, DKFZP564L0562 | |
ANAPC10 | |
IgG | |
This product is specific to Subunit or Isoform: 10. |
Western Blot | |
Unconjugated | |
RUO | |
Q9UM13 | |
10393 | |
Synthetic peptides corresponding to ANAPC10(anaphase promoting complex subunit 10) The peptide sequence was selected from the N terminal of ANAPC10. Peptide sequence MTTPNKTPPGADPKQLERTGTVREIGSQAVWSLSSCKPGFGVDQLRDDNL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title