Learn More
Description
Specifications
Specifications
| Antigen | Apc11 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 85 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | Antiserum with 0.01% Sodium Azide |
| Gene Accession No. | Q9NYG5 |
| Gene Alias | anaphase promoting complex subunit 11, anaphase promoting complex subunit 11 (yeast APC11 homolog), anaphase-promoting complex subunit 11, APC11 anaphase promoting complex subunit 11 homolog, APC11Hepatocellular carcinoma-associated RING finger protein, Apc11p, Cyclosome subunit 11, HSPC214, MGC882 |
| Gene Symbols | ANAPC11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
