Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB355832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355832 Supplier Novus Biologicals Supplier No. NBP177376PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apc2 Polyclonal antibody specifically detects Apc2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Apc2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias adenomatosis polyposis coli 2, adenomatous polyposis coli like, adenomatous polyposis coli protein 2, APCL
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human APC2. The immunogen is located within amino acids 240 - 290 of APC2. Amino Acid Squence: TEVPTHPEDGTPQPGNSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10297
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.