Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc4 Antibody (CIV1.1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250105AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250105AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250105AH Supplier Novus Biologicals Supplier No. NBP250105AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apc4 Monoclonal specifically detects Apc4 in Human samples. It is validated for Western Blot.

Specifications

Antigen Apc4
Applications Western Blot
Classification Monoclonal
Clone CIV1.1
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias anaphase promoting complex subunit 4, anaphase-promoting complex subunit 4, APC4Cyclosome subunit 4
Gene Symbols ANAPC4
Host Species Mouse
Immunogen C-terminus of APC4
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 29945
Test Specificity CIV1.1 may be used for the affinity purification of the Anaphase Promoting Complex (APC/C) by peptide elution. Suggested Dilution for western blot: 1:500. It is suggested to blot for at least 2 hours (wet transfer) to ensure that APC4 is transferred efficiently.Mass spectrometry was used to confirm that the whole APC/C is co-precipitated. To establish the APC4 specific bands, it is suggested to do an APC4 immunoprecipitation assay showing input, IP and FT and blot for at least 2 hours.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.