Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc4 Antibody (CIV1.1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309882 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309882 Supplier Novus Biologicals Supplier No. NBP250105MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apc4 Monoclonal antibody specifically detects Apc4 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Apc4
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone CIV1.1
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias anaphase promoting complex subunit 4, anaphase-promoting complex subunit 4, APC4Cyclosome subunit 4
Host Species Mouse
Immunogen C-terminus of Apc4
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 29945
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.