Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc5 Antibody (APC5#4), HRP, Novus Biologicals™
SDP

Catalog No. NBP250106H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250106H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250106H Supplier Novus Biologicals Supplier No. NBP250106H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apc5 Monoclonal specifically detects Apc5 in Human samples. It is validated for Western Blot.

Specifications

Antigen Apc5
Applications Western Blot
Classification Monoclonal
Clone APC5#4
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5
Gene Symbols ANAPC5
Host Species Mouse
Immunogen GST-human APC5
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51433
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.