Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10055319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055319 Supplier Novus Biologicals Supplier No. NB10055319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Apc5 Polyclonal specifically detects Apc5 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Apc5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2500, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_057321.2
Gene Alias anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5
Gene Symbols ANAPC5
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 705 and the C-terminus (residue 755) of human anaphase promoting complex subunit 5 using the numbering given in entry NP_057321.2 (GeneID 51433).
Molecular Weight of Antigen 85 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51433
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.