Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1 Antibody (2584B) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB401491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401491 Supplier Novus Biologicals Supplier No. FAB10501Z
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

APCDD1 Monoclonal antibody specifically detects APCDD1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen APCDD1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2584B
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human APCDD1, Leu27-His492, Accession # Q8J025
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 147495
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.