Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB11092756C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11092756C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11092756C Supplier Novus Biologicals Supplier No. NB11092756C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APCDD1 Polyclonal specifically detects APCDD1 in Human, Mouse, Chicken, Reptile samples. It is validated for Western Blot.

Specifications

Antigen APCDD1
Applications KnockDown
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q8J025
Gene Alias adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1
Gene Symbols APCDD1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-Prot# Q8J025]
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 147495
Test Specificity Human, mouse, chicken and turtle.
Target Species Human, Mouse, Chicken, Reptile
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.