Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1L Antibody (Z35P1F10*C4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250107AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250107AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250107AH Supplier Novus Biologicals Supplier No. NBP250107AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APCDD1L Monoclonal specifically detects APCDD1L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen APCDD1L
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z35P1F10*C4
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Adenomatosis polyposis coli down-regulated 1 protein-like, adenomatosis polyposis coli down-regulated 1-like, FLJ90166, MGC126807, MGC126809, protein APCDD1-like, RP4-685L9.2
Gene Symbols APCDD1L
Host Species Mouse
Immunogen Peptide Sequence -KRPTSYQA (immunogen peptide is identical in sheep, bovine, porcine, equine and chicken)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 164284
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.