Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1L Antibody (Z35P1F10*C4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395552 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395552 Supplier Novus Biologicals Supplier No. NBP2501070.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APCDD1L Monoclonal antibody specifically detects APCDD1L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen APCDD1L
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z35P1F10*C4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias Adenomatosis polyposis coli down-regulated 1 protein-like, adenomatosis polyposis coli down-regulated 1-like, FLJ90166, MGC126807, MGC126809, protein APCDD1-like, RP4-685L9.2
Host Species Mouse
Immunogen Peptide Sequence -KRPTSYQA (immunogen peptide is identical in sheep, bovine, porcine, equine and chicken)
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 164284
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.