Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APE Antibody (13B8E5C2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB003417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003417 Supplier Novus Biologicals Supplier No. NB100116AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

APE Monoclonal specifically detects APE in Human, Mouse, Primate, Rat samples. It is validated for ChIP Assay, ELISA, Gel Super Shift Assays, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, Western Blot.

Specifications

Antigen APE
Applications ChIP Assay, ELISA, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, Western Blot
Classification Monoclonal
Clone 13B8E5C2
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Gene Symbols APEX1
Host Species Mouse
Immunogen Purified human APE1 [Uniprot: P27695]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.