Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APE Antibody (13B8E5C2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100116SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100116SS 0.025 mg
NB100116 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100116SS Supplier Novus Biologicals Supplier No. NB100116SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 96 publications

APE Monoclonal specifically detects APE in Human, Mouse, Primate, Rat samples. It is validated for Chemotaxis, ChIP Assay, ELISA, EMSA, Gel Super Shift Assays, Gel Supershift, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, Simple Western, Western Blot.

Specifications

Antigen APE
Applications Chemotaxis, ChIP Assay, ELISA, EMSA, Gel Supershift, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, Simple Western
Classification Monoclonal
Clone 13B8E5C2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Simple Western 1:25, ELISA reported in scientific literature (PMID 21769563), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 35286386), Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500, Immunoblotting reported in scientific literature (PMID 27608656), Gel Super Shift Assays reported in scientific literature, Proximity Ligation Assay reported in scientific literature (PMID 27808278), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated
Gene Accession No. P27695
Gene Alias AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Gene Symbols APEX1
Host Species Mouse
Immunogen Purified human APE1 [Uniprot: P27695]
Molecular Weight of Antigen 37 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 328
Target Species Human, Mouse, Mouse, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.